accessibility menu, dialog, popup

UserName

CAS RN 1131-01-7

IUPAC Name: 2-chloro-N-(2,6-dimethylphenyl)acetamide
Synonyms: 2-chloro-N-(2,6-dimethylphenyl)acetamide
Molecular Weight (g/mol): 197.66
Molecular Formula: C10H12ClNO
InChi Key: FPQQSNUTBWFFLB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)CCl
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)