accessibility menu, dialog, popup

UserName

H3CFN

CAS RN 1128-61-6

IUPAC Name: 6-fluoro-2-methylquinoline
Synonyms: 6-fluoro-2-methylquinoline
Molecular Weight (g/mol): 161.18
Molecular Formula: C10H8FN
InChi Key: GPIARMSVZOEZCV-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(F)C=CC2=N1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)