missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-methylquinoline, 98+%

Catalog No. AAA1146922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1146922 100 g
AAA1146906 5 g
AAA1146914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1146922 Supplier Thermo Scientific Chemicals Supplier No. A1146922

CAS: 1128-61-6 | C10H8FN | 161.179 g/mol

Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Bronsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metal-free transfer hydrogenation of 6-fluoro-2-methylquinoline by employing several chiral Brønsted acid catalysts was carried out in combination with dihydropyridines as the hydride source in asymmetric metal-free synthesis of fluoroquinolones by organocatalytic hydrogenation. Hydrogenation product of 6-fluoro-2-methylquinoline is the key intermediate of the antibacterial agent.

Solubility
Soluble in Chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1128-61-6
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.179
MDL Number MFCD00041233
InChI Key GPIARMSVZOEZCV-UHFFFAOYSA-N
Synonym 6-fluoroquinaldine, quinoline, 6-fluoro-2-methyl, 2-methyl-6-fluoroquinoline, 6-fluor-2-methylchinolin, 6-fluoro quinaldine, pubchem7579, acmc-1boyk, maybridge1_000571, 6-fluoro-2-methyl-quinoline, 6-fluoro-2-methyl quinoline
PubChem CID 70784
IUPAC Name 6-fluoro-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)F

Specifications

Melting Point 52°C to 56°C
Quantity 100 g
Beilstein 1281677
Solubility Information Soluble in Chloroform.
Formula Weight 161.18
Percent Purity ≥98%
Chemical Name or Material 6-Fluoro-2-methylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-439-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only