missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-2-methylquinoline 98.0+%, TCI America™

Catalog No. F03005G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. F03005G missing translation for 'mfr' TCI America missing translation for 'supplierNo' F03005G

Chemical Identifiers

CAS 1128-61-6
Molecular Formula C10H8FN
Molecular Weight (g/mol) 161.179
MDL Number MFCD00041233
InChI Key GPIARMSVZOEZCV-UHFFFAOYSA-N
Synonym 6-fluoroquinaldine, quinoline, 6-fluoro-2-methyl, 2-methyl-6-fluoroquinoline, 6-fluor-2-methylchinolin, 6-fluoro quinaldine, pubchem7579, acmc-1boyk, maybridge1_000571, 6-fluoro-2-methyl-quinoline, 6-fluoro-2-methyl quinoline
PubChem CID 70784
IUPAC Name 6-fluoro-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)F

Specifications

Melting Point 53°C
Boiling Point 99°C
Quantity 5 g
Formula Weight 161.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluoro-2-methylquinoline
TSCA No
Recommended Storage Refrigerator