accessibility menu, dialog, popup

UserName

ClH2NNHON

CAS RN 1126-58-5

IUPAC Name: 1-[(hydrazinecarbonyl)methyl]pyridin-1-ium chloride
Synonyms: 1-[(hydrazinecarbonyl)methyl]pyridin-1-ium chloride
Molecular Weight (g/mol): 187.63
Molecular Formula: C7H10ClN3O
InChi Key: NDXLVXDHVHWYFR-UHFFFAOYSA-N
SMILES: [Cl-].NNC(=O)C[N+]1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results