accessibility menu, dialog, popup

UserName

CH3ONH3CNOO

CAS RN 1123-49-5

IUPAC Name: 3,5-dimethyl-4-nitro-1,2-oxazole
Synonyms: 3,5-dimethyl-4-nitro-1,2-oxazole
Molecular Weight (g/mol): 142.11
Molecular Formula: C5H6N2O3
InChi Key: PMQFLWLYAXYFHG-UHFFFAOYSA-N
SMILES: CC1=C(C(C)=NO1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results