missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-nitroisoxazole 98.0+%, TCI America™

Catalog No. D45065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45065G 5 g
D450625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45065G Supplier TCI America Supplier No. D45065G

Chemical Identifiers

CAS 1123-49-5
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.11
MDL Number MFCD00051656
InChI Key PMQFLWLYAXYFHG-UHFFFAOYSA-N
PubChem CID 261410
IUPAC Name 3,5-dimethyl-4-nitro-1,2-oxazole
SMILES CC1=C(C(C)=NO1)[N+]([O-])=O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 142.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-4-nitroisoxazole