accessibility menu, dialog, popup

UserName

H3CNCH3NH

CAS RN 1122-58-3

IUPAC Name: 4-(dimethylamino)pyridin-1-ium
Synonyms: 4-dimethylaminopyridine
Molecular Weight (g/mol): 123.18
Molecular Formula: C7H11N2
InChi Key: VHYFNPMBLIVWCW-UHFFFAOYSA-O
SMILES: CN(C)C1=CC=[NH+]C=C1
Molecular Weight (g/mol) 
  • (4)
  • (10)
  • (1)
Quantity 
  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (6)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (10)
Physical Form 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (1)
  • (3)
  • (11)
  • (4)
  • (10)
  • (1)
  • (5)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (6)
  • (4)
  • (3)
  • (3)
  • (10)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)