accessibility menu, dialog, popup

UserName

CAS RN 1121-89-7

IUPAC Name: piperidine-2,6-dione
Synonyms: glutarimide
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: KNCYXPMJDCCGSJ-UHFFFAOYSA-N
SMILES: O=C1CCCC(=O)N1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)