accessibility menu, dialog, popup

UserName

FI

CAS RN 1121-86-4

IUPAC Name: 1-fluoro-3-iodobenzene
Synonyms: 1-fluoro-3-iodobenzene
Molecular Weight (g/mol): 222
Molecular Formula: C6H4FI
InChi Key: VSKSBSORLCDRHS-UHFFFAOYSA-N
SMILES: FC1=CC=CC(I)=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)