accessibility menu, dialog, popup

UserName

O

CAS RN 1121-66-0

IUPAC Name: cyclohept-2-en-1-one
Synonyms: cyclohept-2-en-1-one
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: WZCRDVTWUYLPTR-UHFFFAOYSA-N
SMILES: O=C1CCCCC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)