accessibility menu, dialog, popup

UserName

H3CO

CAS RN 112-54-9

IUPAC Name: dodecanal
Synonyms: dodecanal duodecylic aldehyde aldehyde C-12, lauric lauric aldehyde C-12 lauric C-12 aldehyde C12 aldehyde lauric aldehyde (natural) laurylaldehyde C-12 aldehyde, lauric
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: HFJRKMMYBMWEAD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC=O
Boiling Point 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)