missing translation for 'onlineSavingsMsg'
Learn More

Dodecanal (stabilized with DL-alpha-Tocopherol) 95.0+%, TCI America™

Catalog No. D0979500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0979500ML 500 mL
D097925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0979500ML Supplier TCI America Supplier No. D0979500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-54-9
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00007017
InChI Key HFJRKMMYBMWEAD-UHFFFAOYSA-N
Synonym lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde
PubChem CID 8194
ChEBI CHEBI:27836
IUPAC Name dodecanal
SMILES CCCCCCCCCCCC=O

Specifications

Color Yellow
Boiling Point 241°C
Quantity 500 mL
Formula Weight 184.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecanal (stabilized with DL-alpha-Tocopherol)
EINECSNumber (2)-0494
RTECSNumber JR1910000
TSCA Yes