missing translation for 'onlineSavingsMsg'
Learn More

Dodecanal, 95%, stab.

Catalog No. AAA1561136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561136 500 g
AAA1561122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1561136 Supplier Thermo Scientific Chemicals Supplier No. A1561136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Store cold | CAS: 112-54-9 | C12H24O | 184.32 g/mol

Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It's end applications include soap, detergent, beauty care product, household product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It′s end applications include soap, detergent, beauty care product, household product.

Solubility
Soluble in chloroform, methanol, alcohol, and propylene glycol. Insoluble in glycerol and water.

Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.

Chemical Identifiers

CAS 112-54-9
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00007017
InChI Key HFJRKMMYBMWEAD-UHFFFAOYSA-N
Synonym lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde
PubChem CID 8194
ChEBI CHEBI:27836
IUPAC Name dodecanal
SMILES CCCCCCCCCCCC=O

Specifications

Melting Point 12°C
Density 0.835
Boiling Point 184°C to 186°C (100 mmHg)
Flash Point 101°C (214°F)
Assay Percent Range 95%
Refractive Index 1.435
Quantity 500 g
Beilstein 1703917
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,methanol,alcohol,and propylene glycol. Insoluble in glycerol and water.
Formula Weight 184.32
Percent Purity 95%
Chemical Name or Material Dodecanal, stabilized with 250ppm BHT
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-983-6
RTECSNumber JR9100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only