missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selectophore™ 1-Dodecanol, ≥98.0%, MilliporeSigma™ Supelco™
Specifications
Specifications
| Melting Point | 22°C to 26°C |
| Density | 0.833 g/mL (at 25°C) |
| Boiling Point | 260°C to 262°C |
| Flash Point | 121°C |
| Ignition Point | 500°F |
| Linear Formula | CH3(CH2)11 OH |
| Quantity | 25 g |
| Vapor Density | 7.4 (vs. air) |
| Formula Weight | 186.33 |
| Vapor Pressure | 0.1 mmHg (20°C) |
| Show More |
Safety and Handling
| RTECSNumber | JR5775000 |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH