accessibility menu, dialog, popup

UserName

CAS RN 112-52-7

H3CCl

CAS RN 112-52-7

IUPAC Name: 1-chlorododecane
Synonyms: 1-chlorododecane
Molecular Weight (g/mol): 204.78
Molecular Formula: C12H25Cl
InChi Key: YAYNEUUHHLGGAH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)