missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethyl ether, 99%

Catalog No. AAL0412822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0412822 100 g
AAL0412836 500 g
AAL041280E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0412822 Supplier Thermo Scientific Chemicals Supplier No. L0412822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 112-49-2 | C8H18O4 | 178.228 g/mol

Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

Solubility
Miscible with water, chloroform and methanol

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.228
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Melting Point -45°C
Density 0.986
Boiling Point 215°C to 216°C
Flash Point 113°C (235°F)
Refractive Index 1.423
Linear Formula CH3O(CH2CH2O)3CH3
Quantity 100 g
Beilstein 1700630
Sensitivity Hygroscopic
Merck Index 14,9692
Solubility Information Miscible with water,chloroform and methanol
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Triethylene glycol dimethyl ether
Show More Show Less
Hazard Category H319-H360Df
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 203-977-3
RTECSNumber XF0665000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only