accessibility menu, dialog, popup

UserName

CAS RN 112-07-2

IUPAC Name: 2-butoxyethyl acetate
Synonyms: butoxyethyl acetate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: NQBXSWAWVZHKBZ-UHFFFAOYSA-N
SMILES: CCCCOCCOC(C)=O
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
  • (8)
  • (3)