accessibility menu, dialog, popup

UserName

NH2H2NOOHHClHCl

CAS RN 11138-66-2

IUPAC Name: 2-(2,4-diaminophenoxy)ethan-1-ol dihydrochloride
Synonyms: 2-(2,4-diaminophenoxy)ethanol dihydrochloride
Molecular Weight (g/mol): 241.11
Molecular Formula: C8H14Cl2N2O2
InChi Key: VXYWXJXCQSDNHX-UHFFFAOYSA-N
SMILES: Cl.Cl.NC1=CC(N)=C(OCCO)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (4)
  • (1)
Percent Purity 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (1)