accessibility menu, dialog, popup

UserName

CAS RN 111128-12-2

IUPAC Name: (2S)-2-[4-(bromomethyl)phenyl]propanoate
Synonyms: (2S)-2-[4-(bromomethyl)phenyl]propanoate
Molecular Weight (g/mol): 242.09
Molecular Formula: C10H10BrO2
InChi Key: QQXBRVQJMKBAOZ-ZETCQYMHSA-M
SMILES: C[C@H](C([O-])=O)C1=CC=C(CBr)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)