missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Bromomethyl)phenyl]propionic acid, 96%

Catalog No. AAH6076314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6076314 25 g
AAH6076306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6076314 Supplier Thermo Scientific Chemicals Supplier No. H6076314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111128-12-2 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111128-12-2
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD02093445
InChI Key QQXBRVQJMKBAOZ-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
PubChem CID 2733978
IUPAC Name 2-[4-(bromomethyl)phenyl]propanoic acid
SMILES CC(C1=CC=C(C=C1)CBr)C(=O)O

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
UN Number UN3261
Formula Weight 243.1
Percent Purity 96%
Chemical Name or Material 2-[4-(Bromomethyl)phenyl]propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only