missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Bromomethyl)phenyl]propionic Acid 98.0+%, TCI America™

Catalog No. B21015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21015G 5 g
B210125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21015G Supplier TCI America Supplier No. B21015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111128-12-2
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD02093445
InChI Key QQXBRVQJMKBAOZ-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
PubChem CID 2733978
IUPAC Name 2-[4-(bromomethyl)phenyl]propanoic acid
SMILES CC(C1=CC=C(C=C1)CBr)C(=O)O

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 243.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[4-(Bromomethyl)phenyl]propionic Acid