accessibility menu, dialog, popup

UserName

CAS RN 111-86-4

IUPAC Name: octan-1-amine
Synonyms: octylamine caprylylamine armeen 8D caprylamine armeen 8
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: IOQPZZOEVPZRBK-UHFFFAOYSA-N
SMILES: CCCCCCCCN
Boiling Point 
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)