missing translation for 'onlineSavingsMsg'
Learn More

1-Octylamine, 99%

Catalog No. AAB2419336
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2419336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2419336 Supplier Thermo Scientific Chemicals Supplier No. B2419336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 111-86-4 | C8H19N | 129.247 g/mol

1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.

Chemical Identifiers

CAS 111-86-4
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008247
InChI Key IOQPZZOEVPZRBK-UHFFFAOYSA-N
Synonym octylamine, n-octylamine, 1-aminooctane, 1-octanamine, 1-octylamine, caprylamine, caprylylamine, armeen 8, n-octylamine, mono, armeen 8d
PubChem CID 8143
ChEBI CHEBI:7728
IUPAC Name octan-1-amine
SMILES CCCCCCCCN

Specifications

Melting Point -1°C
Density 0.78
Boiling Point 178°C to 179°C
Flash Point 60°C (144°F)
Refractive Index 1.429
Quantity 500 g
UN Number UN2735
Beilstein 1679227
Sensitivity Air Sensitive
Solubility Information Miscible with ethanol,benzene and ethyl acetate. Slightly miscible with water.
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material 1-Octylamine
Show More Show Less
Hazard Category H226-H301+H311-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 203-916-0
RTECSNumber RG8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only