accessibility menu, dialog, popup

UserName

CAS RN 111-78-4

IUPAC Name: (1Z,5Z)-cycloocta-1,5-diene
Synonyms: 1,5-cyclooctadiene, (Z,Z)-
Molecular Weight (g/mol): 108.18
Molecular Formula: C8H12
InChi Key: VYXHVRARDIDEHS-QGTKBVGQSA-N
SMILES: C1C\C=C/CC\C=C/1
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)