accessibility menu, dialog, popup

UserName

H3COOOH

CAS RN 111-77-3

IUPAC Name: 2-(2-methoxyethoxy)ethan-1-ol
Synonyms: diglycol methyl ether
Molecular Weight (g/mol): 120.15
Molecular Formula: C5H12O3
InChi Key: SBASXUCJHJRPEV-UHFFFAOYSA-N
SMILES: COCCOCCO
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (8)
  • (3)