accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 111-50-2

IUPAC Name: hexanedioyl dichloride
Synonyms: adipoyl chloride
Molecular Weight (g/mol): 183.03
Molecular Formula: C6H8Cl2O2
InChi Key: PWAXUOGZOSVGBO-UHFFFAOYSA-N
SMILES: ClC(=O)CCCCC(Cl)=O
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)