CAS RN 11096-82-5
IUPAC Name:
2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Synonyms:
2,2',3,3',4,4'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol):
360.86
Molecular Formula:
C12H4Cl6
InChi Key:
BTAGRXWGMYTPBY-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C(Cl)=C1Cl)C1=CC=C(Cl)C(Cl)=C1Cl
1
–
2
of
2
results
2
Aroclor 1260, SPEX CertiPrep™
CAS: 11096-82-5 Molecular Formula: C12H4Cl6 Molecular Weight (g/mol): 360.864 InChI Key: BTAGRXWGMYTPBY-UHFFFAOYSA-N PubChem CID: 38018 IUPAC Name: 1,2,3-trichloro-4-(2,3,4-trichlorophenyl)benzene SMILES: C1=CC(=C(C(=C1C2=C(C(=C(C=C2)Cl)Cl)Cl)Cl)Cl)Cl