missing translation for 'onlineSavingsMsg'
Learn More

Aroclor 1260 Stock Solution, 1000μg/mL in methanol, Ultra Scientific

Catalog No. USPPS1411
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPPS1411 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPPS1411 Supplier Agilent BioTek Supplier No. PPS1411

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Aroclor 1260 Stock Solution
CAS 11096-82-5, 67-56-1
CAS Max % 99.87, 0.11
Analyte or Component Names Methanol,Aroclor 1260
Element Methanol, Aroclor 1260
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL