accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OH2OCH3OO(S)(R)OHOH(S)(R)HOCH3(R)HOHOOOHOHOHO

CAS RN 11018-89-6

IUPAC Name: 4-[(1R,7S,9R,9bS,10R)-3a,5a,9,10-tetrahydroxy-9a-(hydroxymethyl)-11a-methyl-7-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one octahydrate
Synonyms: 4-[(1R,7S,9R,9bS,10R)-3a,5a,9,10-tetrahydroxy-9a-(hydroxymethyl)-11a-methyl-7-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]-dodecahydro-1H-cyclopenta[a]phenanthren-1-yl]-5H-furan-2-one octahydrate
Molecular Weight (g/mol): 728.78
Molecular Formula: C29H60O20
InChi Key: TYBARJRCFHUHSN-HMNHGHQSSA-N
SMILES: O.O.O.O.O.O.O.O.CC1OC(O[C@H]2C[C@@H](O)C3(CO)[C@H]4[C@H](O)CC5(C)[C@H](CCC5(O)C4CCC3(O)C2)C2=CC(=O)OC2)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)