missing translation for 'onlineSavingsMsg'
Learn More

Ouabain octahydrate, 95%

Catalog No. AAJ6072403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6072403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6072403 Supplier Thermo Scientific Chemicals Supplier No. J6072403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of sodium-potassium-dependent ATPase | CAS: 11018-89-6 | C29H60O20 | 728.779 g/mol

Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ouabain, as well as other endogenous digitalis-like compounds, can down-regulate the expression of 14-3-3 proteins in rat lens.

Solubility
Soluble in water.

Notes
Protect from light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Specifications

CAS 11018-89-6
Color White
Molecular Formula C29H60O20
Quantity 1 g
MDL Number MFCD00149240
UN Number UN1544
Beilstein 101712
Sensitivity Light sensitive
Merck Index 14,6901
Synonym ouabain octahydrate, strophantine octahydrate, quabain octahydrate, ouabain usp, penta a phenanthren-17-yl-2h-furan-5-one;octahydrate, gamma-strophanthin, 3-1r,3s,5s,8r,9s,10r,11r,13r,14s,17r-1,5,11,14-tetrahydroxy-10-hydroxymethyl-13-methyl-3-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyloxan-2-yl oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1h-cyclo, prestwick_370, acocantherine;g-strophanthin
Solubility Information Soluble in water.
InChI Key TYBARJRCFHUHSN-DMJRSANLSA-N
SMILES CC1C(C(C(C(O1)OC2CC(C3(C4C(CCC3(C2)O)C5(CCC(C5(CC4O)C)C6=CC(=O)OC6)O)CO)O)O)O)O.O.O.O.O.O.O.O.O
IUPAC Name 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one;octahydrate
Molecular Weight (g/mol) 728.779
PubChem CID 6364534
Formula Weight 728.78 (584.66 Anhydrous)
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ouabain octahydrate
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
RTECSNumber RN3675000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only