CAS RN 110117-83-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ 1-Methyl-D-tryptophan
Indoleamine 2,3-dioxygenase (IDO) inhibitor