accessibility menu, dialog, popup

UserName

ClOCH2

CAS RN 110-75-8

IUPAC Name: (2-chloroethoxy)ethene
Synonyms: 2-chloroethyl vinyl ether
Molecular Weight (g/mol): 106.55
Molecular Formula: C4H7ClO
InChi Key: DNJRKFKAFWSXSE-UHFFFAOYSA-N
SMILES: ClCCOC=C
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)