missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine) 97.0+%, TCI America™

Catalog No. C0174500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0174500ML 500 mL
C017425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0174500ML Supplier TCI America Supplier No. C0174500ML
Only null left

Chemical Identifiers

CAS 110-75-8
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00000973
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
Synonym 2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether
PubChem CID 8074
IUPAC Name 1-chloro-2-ethenoxyethane
SMILES C=COCCCl

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 108°C
Quantity 500 mL
UN Number 1992
Formula Weight 106.55
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine)
EINECSNumber (2)-0377
RTECSNumber KN6300000
TSCA Yes