missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Ethylamino)ethanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -90°C |
| Color | Yellow |
| Boiling Point | 169°C |
| Quantity | 500 mL |
| UN Number | 2922 |
| Formula Weight | 89.14 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(Ethylamino)ethanol |
Safety and Handling
| EINECSNumber | (2)-0295 |
| RTECSNumber | KK9100000 |
| TSCA | Yes |