missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%

Catalog No. AAA15744AP
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15744AP 500 mL
AAA157440F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA15744AP Supplier Thermo Scientific Chemicals Supplier No. A15744AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-73-6 | C4H11NO | 89.138 g/mol

2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Ethylamino)ethanol is an efficient compound used for inhibiting corrosion. This high activity of the inhibitor may be interpreted both by its ability to stabilize the hydroxide surface film and by its adsorption onto bare sites of the metal.

Solubility
Miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO

Specifications

Melting Point -9°C
Density 0.914
Boiling Point 168°C to 170°C
Flash Point 71°C (160°F)
Refractive Index 1.441
Linear Formula CH3CH2NHCH2CH2OH
Quantity 500 mL
UN Number UN2922
Beilstein 635671
Solubility Information Miscible in water.
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material 2-(Ethylamino)ethanol
Show More Show Less
Hazard Category H227-H302-H311-H314
Hazard Statement GHS H Statement
H311-H314-H318-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-797-5
RTECSNumber KK9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only