accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 110-71-4

IUPAC Name: 1,2-dimethoxyethane
Synonyms: 1,2-dimethoxyethane ethylenglykoldimethylether hisolve mmm DME dme (glycol ether) dimethyl cellosolve ansul ether 121 dme 1,2-ethanediol dimethyl ether ethylene dimethyl ether
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: XTHFKEDIFFGKHM-UHFFFAOYSA-N
SMILES: COCCOC
Boiling Point 
  • (7)
  • (2)
  • (3)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (7)
  • (9)
Percent Purity 
  • (6)
  • (2)
  • (1)
  • (7)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (13)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (7)
  • (1)
  • (3)
  • (7)
  • (2)
  • (3)
  • (7)
  • (2)
  • (3)
  • (1)
  • (3)
  • (7)
  • (9)
  • (6)
  • (2)
  • (1)
  • (7)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (13)
  • (1)
  • (1)
  • (7)