missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dimethoxyethane, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018779
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018779 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018779 Supplier Merck Emd Millipore Supplier No. 724055MLF

1,2-Dimethoxyethane is a water miscible solvent, used as an alternative for the hazardous organic solvents such as dioxane and tetrahydrofuran.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 396°F
CAS 110-71-4
Vapor Pressure 48 mmHg (20°C)
Vapor Density 3.1 (20°C, vs. air)
Density 0.867 g/mL (at 25°C)
Flash Point -2°C
Physical Form Neat
Linear Formula CH3 OCH2 CH2 OCH3
MDL Number MFCD00008502
Melting Point -58°C
Boiling Point 85°C
Synonym mono-Glyme; Dimethylglycol; Ethylene glycol dimethyl ether; Monoglyme
Type Analytical Standard
Quantity 5 mL
Formula Weight 90.12
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less