accessibility menu, dialog, popup

UserName

CAS RN 1098102-95-4

H3COSSSnCH3CH3H3COCH3SnH3CCH3H3C

CAS RN 1098102-95-4

IUPAC Name: [2,8-bis(octyloxy)-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl]trimethylstannane
Synonyms: [2,8-bis(octyloxy)-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl]trimethylstannane
Molecular Weight (g/mol): 772.32
Molecular Formula: C32H54O2S2Sn2
InChi Key: CDHNVAGJGMNTLW-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C2C=C(SC2=C(OCCCCCCCC)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results