missing translation for 'onlineSavingsMsg'
Learn More

4,8-Bis(n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene 95.0+%, TCI America™

Catalog No. B4378200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4378200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4378200MG Supplier TCI America Supplier No. B4378200MG
Only null left

Chemical Identifiers

CAS 1098102-95-4
Molecular Formula C32H54O2S2Sn2
Molecular Weight (g/mol) 772.32
MDL Number MFCD27923034
InChI Key CDHNVAGJGMNTLW-UHFFFAOYSA-N
PubChem CID 71721478
IUPAC Name [2,8-bis(octyloxy)-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl]trimethylstannane
SMILES CCCCCCCCOC1=C2C=C(SC2=C(OCCCCCCCC)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C

Specifications

Quantity 200 mg
UN Number 3146
Formula Weight 772.32
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis(n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b′]dithiophene