accessibility menu, dialog, popup

UserName

CAS RN 109-74-0

IUPAC Name: butanenitrile
Synonyms: butyronitrile butyric acid nitrile
Molecular Weight (g/mol): 69.11
Molecular Formula: C4H7N
InChi Key: KVNRLNFWIYMESJ-UHFFFAOYSA-N
SMILES: CCCC#N
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)