missing translation for 'onlineSavingsMsg'
Learn More

n-Butyronitrile, 99%

Catalog No. AC108145000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC108145000 500 mL
AC108141000 100 mL
AC108140025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108145000 Supplier Thermo Scientific Chemicals Supplier No. 108145000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-74-0 | C4H7N | 69.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-74-0
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -112.0°C
Density 0.7900g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7N
Refractive Index 1.3830 to 1.3850
Linear Formula CH3CH2CH2CN
Quantity 500 mL
Beilstein 02, II, 252
Specific Gravity 0.79
Merck Index 15, 1601
Solubility Information Solubility in water: 33g/L (25°C). Other solubilities: miscible with alcohol,ether and dimethylformamide
Molecular Weight (g/mol) 69.11
Viscosity 0.57 mPa.s (20°C)
Formula Weight 69.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyronitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-700-6

RUO – Research Use Only