missing translation for 'onlineSavingsMsg'
Learn More

Butyronitrile, 99%

Catalog No. AAL02999AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL02999AU 1000 mL
AAL02999AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL02999AU Supplier Thermo Scientific Chemicals Supplier No. L02999AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 109-74-0 | C4H7N | 69.107 g/mol

Butyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Butyronitrile serves as an intermediate in the chemical and pharmaceutical industries. It is also involved in the preparation of n-butylamine, butanamide and butyric acid. It acts as a precursor to the poultry drug amprolium. Further, it reacts with phosphorochloridic acid diethyl ester to prepare (1-cyano-propyl)-phosphonic acid diethyl ester. It is also employed to study the effect of stereochemical factors on intervalence charge transfer. In addition to this, it is used in electrolyte composition in dye-sensitized solar cells.

Solubility
Miscible with benzene, alcohol, ether and dimethylformamide. Slightly soluble in water.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 109-74-0
Molecular Formula C4H7N
Molecular Weight (g/mol) 69.107
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Specifications

Melting Point -112°C
Density 0.792
Boiling Point 115°C to 117°C
Flash Point 18°C (64°F)
Refractive Index 1.384
Linear Formula CH3CH2CH2C≡N
Quantity 1000 mL
UN Number UN2411
Beilstein 1361452
Merck Index 14,1597
Solubility Information Miscible with benzene,alcohol,ether and dimethylformamide. Slightly soluble in water.
Formula Weight 69.11
Percent Purity 99%
Chemical Name or Material Butyronitrile
Show More Show Less
Hazard Category H225-H300-H311+H331
Hazard Statement GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYRONITRILE
EINECSNumber 203-700-6
RTECSNumber ET8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only