CAS RN 108341-18-0
IUPAC Name:
(2R,3R)-2,3-dihydroxybutanedioic acid 4-[(1R)-2-amino-1-hydroxyethyl]benzene-1,2-diol hydrate
Synonyms:
L(+)-tartaric acid norepinephrine hydrate
Molecular Weight (g/mol):
337.28
Molecular Formula:
C12H19NO10
InChi Key:
LNBCGLZYLJMGKP-LUDZCAPTSA-N
SMILES:
O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@H](O)C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
L-Noradrenaline Bitartrate Monohydrate 98.0+%, TCI America™
CAS: 108341-18-0 Molecular Formula: C12H19NO10 Molecular Weight (g/mol): 337.281 MDL Number: MFCD00036384 InChI Key: LNBCGLZYLJMGKP-PYYOGZMQSA-N Synonym: L-Arterenol Bitartrate, L-Norepinephrine Bitartrate PubChem CID: 71311448 IUPAC Name: 4-[(1S)-2-amino-1-hydroxyethyl]benzene-1,2-diol;(2R,3R)-2,3-dihydroxybutanedioic acid;hydrate SMILES: C1=CC(=C(C=C1C(CN)O)O)O.C(C(C(=O)O)O)(C(=O)O)O.O