accessibility menu, dialog, popup

UserName

CAS RN 108124-17-0

IUPAC Name: 2-methyl-5-phenylfuran-3-carboxylate
Synonyms: 2-methyl-5-phenylfuran-3-carboxylate
Molecular Weight (g/mol): 201.2
Molecular Formula: C12H9O3
InChi Key: VLMNACSEESRUAK-UHFFFAOYSA-M
SMILES: CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results