missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furoic acid, 97%, Thermo Scientific™

Catalog No. CC14901DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC14901DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC14901DA Supplier Thermo Scientific Chemicals Supplier No. CC14901DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108124-17-0
Molecular Formula C12H9O3
Molecular Weight (g/mol) 201.20
MDL Number MFCD00221070
InChI Key VLMNACSEESRUAK-UHFFFAOYSA-M
Synonym 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid
PubChem CID 736516
IUPAC Name 2-methyl-5-phenylfuran-3-carboxylic acid
SMILES CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-phenyl-3-furoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.