missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furoic acid, 97%

Catalog No. AAB2098806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2098806 5 g
AAB2098803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2098806 Supplier Thermo Scientific Chemicals Supplier No. B2098806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108124-17-0 | C12H9O3 | 201.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108124-17-0
Molecular Formula C12H9O3
Molecular Weight (g/mol) 201.20
MDL Number MFCD00221070
InChI Key VLMNACSEESRUAK-UHFFFAOYSA-M
Synonym 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid
PubChem CID 736516
SMILES CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 181°C to 184°C
Quantity 5 g
IUPAC Name 2-methyl-5-phenylfuran-3-carboxylate
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-phenyl-3-furoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only