accessibility menu, dialog, popup

UserName

OHOH

CAS RN 1079-21-6

IUPAC Name: [1,1'-biphenyl]-2,5-diol
Synonyms: phenylhydroquinone
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: XCZKKZXWDBOGPA-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results