accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 107868-30-4

H3C(S)(S)(R)CH2O(R)H3C(S)O

CAS RN 107868-30-4

IUPAC Name: (3aS,3bR,9aR,9bS,11aS)-9a,11a-dimethyl-5-methylidene-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-dione
Synonyms: exemestane pharmacia brand of exemestane pharmacia spain brand of exemestane aromasil aromasin aromasine exemestanum exemestance exemestano [inn-spanish] exemestane (product)
Molecular Weight (g/mol): 296.41
Molecular Formula: C20H24O2
InChi Key: BFYIZQONLCFLEV-DAELLWKTSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CC(=C)C4=CC(=O)C=C[C@]34C)[C@@H]1CCC2=O
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.