missing translation for 'onlineSavingsMsg'
Learn More

Exemestane Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012417
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012417 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012417 Supplier Merck Emd Millipore Supplier No. PHR16341G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Exemestane is a steroidal anticancer drug, that belongs to the class of antiestrogens known as aromatase inhibitors. It is commonly used for the treatment of breast cancer. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 107868-30-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H24O2
MDL Number MFCD00866994
Synonym Exemestane; 6-Methyleneandrosta-1,4-diene-3,17-dione
Type Certified Reference Material
Quantity 1 g
Formula Weight 296.40
Grade Certified Reference Material